Tetrabutylammoniumhexafluorophosphat
Lieferant: Molekula
Synonyme:
Tetrabutylammonium hexafluorophosphate(V), Tetra-n-butylammonium hexafluorophosphate
32102353-100G
MOLE32102353-100GEA
155
EUR
MOLE32102353-100G
Tetrabutylammoniumhexafluorophosphat
Tetrabutylammonium hexafluorophosphate
Formel:
C₁₆H₃₆F₆NP Molecular Weight: 387,43 g/mol |
MDL:
MFCD00011748 CAS-Nummer: 3109-63-5 EINECS: 221-472-6 UN: 3263 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...