Actinomycin D

Lieferant: Molekula

32117981-25MG
MOLE32117981-25MGEA 372 EUR
MOLE32117981-25MG
Actinomycin D
Actinomycin D
Formel: C₆₂H₈₆N₁₂O₁₆
Molecular Weight: 1255,5 g/mol
MDL: MFCD00005033
CAS-Nummer: 50-76-0
UN: 3462
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR