2-Chlor-1-methylpyridiniumiodid
Lieferant: Molekula
Synonyme:
2-Chlorpyridin 1-methoiodid
32135861-100G
MOLE32135861-100GEA
0
EUR
MOLE32135861-100G
2-Chlor-1-methylpyridiniumiodid
2-Chloro-1-methylpyridinium iodide
Formel:
C₆H₇ClIN Molecular Weight: 255,49 g/mol Schmelzpunkt: 207 °C |
MDL:
MFCD00011984 CAS-Nummer: 14338-32-0 EINECS: 238-288-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...