α,α'-Dichlor-m-xylol
Lieferant: Molekula
Synonyme:
m-Xyloldichlorid, 1,3-Bis(chlormethyl)benzol, m-Xylylendichlorid
32404482-100G
MOLE32404482-100GEA
346
EUR
MOLE32404482-100G
α,α'-Dichlor-m-xylol
α,α'-Dichlor-m-xylol
Formel:
C₈H₈Cl₂ Molecular Weight: 175,06 g/mol Siedepunkt: 254 °C (1013 hPa) Schmelzpunkt: 33…35 °C Dichte: 1,302 g/cm³ (20 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00000912 CAS-Nummer: 626-16-4 EINECS: 210-932-1 UN: 2928 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...