(S)-(-)-4-Isopropyl-2-oxazolidinon
Lieferant: Molekula
Synonyme:
(4S)-(-)-Isopropyl-2-oxazolidinone, (4S)-(-)-4-Isopropyl-2-oxazolidinone, (S)-4-Isopropyl-2-oxazolidinone, (4S)-2-Oxo-4-(prop-2-yl)-1,3-oxazolidine, (4S)-4-(Prop-2-yl)-1,3-oxazolidin-2-one
32506253-1G
MOLE32506253-1GEA
172
EUR
MOLE32506253-1G
(S)-(-)-4-Isopropyl-2-oxazolidinon
(S)-(-)-4-Isopropyl-2-oxazolidinon
Formel:
C₆H₁₁NO₂ Molecular Weight: 129,16 g/mol |
MDL:
MFCD00010847 CAS-Nummer: 17016-83-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...