4-Brom-3,5-dimethoxybenzoesäure

Lieferant: Molekula

32596516-25G
MOLE32596516-25GEA 0 EUR
MOLE32596516-25G
4-Brom-3,5-dimethoxybenzoesäure
4-Bromo-3,5-dimethoxybenzoic acid
Formel: C₉H₉BrO₄
Molecular Weight: 261,07 g/mol
MDL: MFCD01632140
CAS-Nummer: 56518-42-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR