(S)-(-)-β-Methylphenethylamin

Lieferant: Molekula

Synonyme: (S)-(-)-2-Phenyl-1-propylamine, (S)-(-)-beta-Methylphenethylamine

32642343-1G
MOLE32642343-1GEA 0 EUR
MOLE32642343-1G
(S)-(-)-β-Methylphenethylamin
(S)-(-)-β-Methylphenethylamin
Formel: C₉H₁₃N
MDL: MFCD00216741
CAS-Nummer: 17596-79-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR