(2R,3R)-(-)-2,3-O-Isopropyliden-D-threitol
Lieferant: Molekula
Synonyme:
(-)-2,3-O-Isopropylidene-D-threitol
32713857-1G
MOLE32713857-1GEA
0
EUR
MOLE32713857-1G
(2R,3R)-(-)-2,3-O-Isopropyliden-D-threitol
(2R,3R)-(-)-2,3-O-Isopropyliden-D-threitol
Formel:
C₇H₁₄O₄ Molecular Weight: 162,18 g/mol |
MDL:
MFCD00009761 CAS-Nummer: 73346-74-4 EINECS: 277-391-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...