(R)-(-)-β-Aminophenethylalkohol
Lieferant: Molekula
Synonyme:
D-Phenylglycinol, (R)-(-)-2-Phenylglycinol
32798842-5G
MOLE32798842-5GEA
0
EUR
MOLE32798842-5G
(R)-(-)-β-Aminophenethylalkohol
(R)-(-)-β-Aminophenethylalkohol
Formel:
C₈H₁₁NO Molecular Weight: 137,18 g/mol |
MDL:
MFCD00008062 CAS-Nummer: 56613-80-0 EINECS: 260-287-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...