4-Brom-m-toluidin

Lieferant: Molekula

32976775-25G
MOLE32976775-25GEA 0 EUR
MOLE32976775-25G
4-Brom-m-toluidin
4-Brom-m-toluidin
Formel: C₇H₈BrN
MDL: MFCD00007828
CAS-Nummer: 6933-10-4
EINECS: 230-056-3
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR