2-Chlorbenzimidazol

Lieferant: Molekula

Synonyme: 2-Chlor-1H-benzimidazol

32987316-25G
MOLE32987316-25GEA 0 EUR
MOLE32987316-25G
2-Chlorbenzimidazol
2-Chlorobenzimidazole
Formel: C₇H₅ClN₂
Molecular Weight: 152.58 g/mol
MDL: MFCD00051944
CAS-Nummer: 4857-06-1
EINECS: 225-453-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR