4-Chlor-2-nitrobenzylalkohol

Lieferant: Molekula

33035622-5G
MOLE33035622-5GEA 0 EUR
MOLE33035622-5G
4-Chlor-2-nitrobenzylalkohol
4-Chlor-2-nitrobenzylalkohol
Formel: C₇H₆ClNO₃
Molecular Weight: 187,58 g/mol
Schmelzpunkt: 89…90 °C
MDL: MFCD00007217
CAS-Nummer: 22996-18-5
EINECS: 245-374-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR