Ethyl-2-chlornicotinat
Lieferant: Molekula
Synonyme:
Ethyl 2-chloropyridine-3-carboxylate, Ethyl 2-Chloro-3-pyridinecarboxylate, Ethyl 2-chloronicotinate
33162912-25G
MOLE33162912-25GEA
0
EUR
MOLE33162912-25G
Ethyl-2-chlornicotinat
Ethyl-2-chlornicotinat
Formel:
C₈H₈ClNO₂ Molecular Weight: 185,61 g/mol |
MDL:
MFCD00674129 CAS-Nummer: 1452-94-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...