2-(3-Chlorpropyl)-1,3-dioxolan

Lieferant: Molekula

Synonyme: 2-(3-chloropropyl)-1,3-dioxolane, 4-Chlorobutyraldehyde ethylene Acetal

33355557-5G
MOLE33355557-5GEA 0 EUR
MOLE33355557-5G
2-(3-Chlorpropyl)-1,3-dioxolan
2-(3-Chlorpropyl)-1,3-dioxolan
Formel: C₆H₁₁ClO₂
Molecular Weight: 150,6 g/mol
MDL: MFCD00043135
CAS-Nummer: 16686-11-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR