2,4-Dimethoxybenzylalkohol

Lieferant: Molekula

33739999-25G
MOLE33739999-25GEA 0 EUR
MOLE33739999-25G
2,4-Dimethoxybenzylalkohol
2,4-Dimethoxybenzylalkohol
Formel: C₉H₁₂O₃
Molecular Weight: 168,19 g/mol
Siedepunkt: 317 °C (1013 hPa)
Schmelzpunkt: 34…35 °C
MDL: MFCD00004614
CAS-Nummer: 7314-44-5
EINECS: 230-775-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR