Bariumtrifluormethansulfonat

Lieferant: Molekula

Synonyme: Barium Bis(trifluoromethanesulphonate), Barium triflate, Barium(II) trifluoromethanesulphonate, Trifluoromethanesulphonic acid Barium(II) salt, Ba(OTf)2, Barium(II) triflate

33858797-5G
MOLE33858797-5GEA 0 EUR
MOLE33858797-5G
Bariumtrifluormethansulfonat
Bariumtrifluormethansulfonat
Formel: C₂BaF₆O₆S₂
Molecular Weight: 435,45 g/mol
MDL: MFCD00143644
CAS-Nummer: 2794-60-7
EINECS: 220-526-6
UN: 1564
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR