N-(Hydroxymethyl)acetamid

Lieferant: Molekula

Synonyme: N-Hydroxymethylacetamide, N-Acetylmethanolamine, Acetamidomethanol

33937183-25G
MOLE33937183-25GEA 0 EUR
MOLE33937183-25G
N-(Hydroxymethyl)acetamid
N-(Hydroxymethyl)acetamid
Formel: C₃H₇NO₂
Molecular Weight: 89,09 g/mol
MDL: MFCD00014417
CAS-Nummer: 625-51-4
EINECS: 210-897-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR