2,4,5-Trimethyloxazol
Lieferant: Molekula
Synonyme:
2,4,5-Trimethyl-1,3-oxazol
34114795-25G
MOLE34114795-25GEA
0
EUR
MOLE34114795-25G
2,4,5-Trimethyloxazol
2,4,5-Trimethyloxazole
Formel:
C₆H₉NO Molecular Weight: 111,14 g/mol Siedepunkt: 133 °C (1013 hPa) Dichte: 0,9569 g/cm³ (20 °C) |
MDL:
MFCD00005308 CAS-Nummer: 20662-84-4 EINECS: 243-952-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...