5-Bromchinolin

Lieferant: Molekula
Achtung

34200144-1G
MOLE34200144-1GEA 71.6 EUR
MOLE34200144-1G
5-Bromchinolin
5-Bromoquinoline
Formel: C₉H₆BrN
Molecular Weight: 208,06 g/mol
MDL: MFCD00234481
CAS-Nummer: 4964-71-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR