DIPSO
Lieferant: Molekula
Synonyme:
3-(Bis(2-hydroxyethyl)amino)-2-hydroxypropane-1-sulphonic acid, DIPSO, 3-[Bis(2-hydroxyethyl)amino]-2-hydroxypropanesulphonic acid
34218155-100G
MOLE34218155-100GEA
131
EUR
MOLE34218155-100G
DIPSO
DIPSO
Formel:
C₇H₁₇NO₆S Molecular Weight: 261.29 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00038353 CAS-Nummer: 68399-80-4 EINECS: 269-992-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...