2-Thiophenethanol
Lieferant: Molekula
Synonyme:
2-(2-Thienyl)ethanol, 2-(2-Hydroxyethyl)thiophen, Thiophen-2-ethanol
34607299-5G
MOLE34607299-5GEA
0
EUR
MOLE34607299-5G
2-Thiophenethanol
2-Thiophenethanol
Formel:
C₆H₈OS Siedepunkt: 108 °C (1013 hPa) Dichte: 1,197 g/cm³ (20 °C) |
MDL:
MFCD00005462 CAS-Nummer: 5402-55-1 EINECS: 226-452-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...