2'-Desoxyadenosin Monohydrat
Lieferant: Molekula
Synonyme:
(2R,3S,5R)-5-(6-Amino-9H-purin-9-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol hydrate
34645670-5G
MOLE34645670-5GEA
96.8
EUR
MOLE34645670-5G
2'-Desoxyadenosin Monohydrat
2'-Desoxyadenosin Monohydrat
Formel:
C₁₀H₁₃N₅O₃·1H₂O Molecular Weight: 269,26 g/mol (251,24 anhy) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149364 CAS-Nummer: 16373-93-6 EINECS: 213-488-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...