(±)-Tetrahydrofurfurylchlorid

Lieferant: Molekula

34755704-100G
MOLE34755704-100GEA 0 EUR
MOLE34755704-100G
(±)-Tetrahydrofurfurylchlorid
(±)-Tetrahydrofurfurylchlorid
Formel: C₅H₉ClO
Molecular Weight: 120,58 g/mol
MDL: MFCD00005370
CAS-Nummer: 3003-84-7
EINECS: 221-102-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR