Natriumdiphenylamin-4-sulfonat

Lieferant: Molekula

Synonyme: 4-(Phenylamino)benzolsulfonsäure Natriumsalz, Diphenylamin-4-sulfonsäure Natriumsalz, Natrium(4-anilinobenzolsulfonat), Natrium-N-phenylsulfanilat

34781230-5G
MOLE34781230-5GEA 101 EUR
MOLE34781230-5G
Natriumdiphenylamin-4-sulfonat
Natriumdiphenylamin-4-sulfonat
Formel: C₁₂H₁₀NNaO₃S
Molecular Weight: 271,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00036468
CAS-Nummer: 6152-67-6
EINECS: 228-165-6
Merck Index: 13,07397

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR