6-Methoxy-1-indanon

Lieferant: Molekula

34858611-5G
MOLE34858611-5GEA 121 EUR
MOLE34858611-5G
6-Methoxy-1-indanon
6-Methoxy-1-indanon
Formel: C₁₀H₁₀O₂
Molecular Weight: 162,19 g/mol
Siedepunkt: 270 °C (1013 hPa)
Schmelzpunkt: 108 °C
MDL: MFCD00021232
CAS-Nummer: 13623-25-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR