(±)-Ethyl-2,3-dibrompropionat

Lieferant: Molekula

Synonyme: Ethyl 2,3-Dibromopropionate

34893089-100G
MOLE34893089-100GEA 0 EUR
MOLE34893089-100G
(±)-Ethyl-2,3-dibrompropionat
(±)-Ethyl-2,3-dibrompropionat
Formel: C₅H₈Br₂O₂
Molecular Weight: 259,93 g/mol
MDL: MFCD00000212
CAS-Nummer: 3674-13-3
EINECS: 222-941-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR