Butyrylthiocholiniodid
Lieferant: Molekula
Synonyme:
Butyrylthiocholiniumiodid, (Propylcarbonylthioethyl)trimethylammoniumiodid, (2-(Butyrylthio)ethyl)trimethylammoniumiodid, (2-Butyrylthioethyl)trimethylammoniumiodid
34904481-5G
MOLE34904481-5GEA
116
EUR
MOLE34904481-5G
Butyrylthiocholiniodid
Butyrylthiocholiniodid
Formel:
H₃C(CH₂)₂C(S)O(CH₂)₂N(CH₃)₃I Molecular Weight: 317,23 g/mol Schmelzpunkt: 172 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00011845 CAS-Nummer: 1866-16-6 EINECS: 217-475-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...