7-Chlorindol

Lieferant: Molekula
Achtung

35064238-1G
MOLE35064238-1GEA 0 EUR
MOLE35064238-1G
7-Chlorindol
7-Chlorindol
Formel: C₈H₆ClN
Molecular Weight: 151,6 g/mol
MDL: MFCD01321301
CAS-Nummer: 53924-05-3
EINECS: 258-865-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR