1-Phenyl-2-tetrazolin-5-thion
Lieferant: Molekula
Synonyme:
1-Phenyl-5-mercaptotetrazol, 5-Mercapto-1-phenyl-1H-tetrazol, 1-Phenyl-1H-tetrazol-5-thiol
35197222-100G
MOLE35197222-100GEA
184
EUR
MOLE35197222-100G
1-Phenyl-2-tetrazolin-5-thion
1-Phenyl-2-tetrazolin-5-thion
Formel:
C₇H₆N₄S Molecular Weight: 178,22 g/mol Schmelzpunkt: 146…150 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003129 CAS-Nummer: 86-93-1 EINECS: 201-710-5 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...