D(-)-α-Phenylglycin

Lieferant: Molekula

Synonyme: D-(-)-2-Phenylglycine, H-D-Phg-OH, (R)-(-)-α-Aminophenylacetic acid

35200423-100G
MOLE35200423-100GEA 146 EUR
MOLE35200423-100G
D(-)-α-Phenylglycin
D(-)-α-Phenylglycin
Formel: C₈H₉NO₂
Molecular Weight: 151,16 g/mol
MDL: MFCD00008061
CAS-Nummer: 875-74-1
EINECS: 212-876-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR