5-Azacytosin
Lieferant: Molekula
Synonyme:
4-Amino-1,3,5-triazin-2(1H)-one, 6-Amino-1,3,5-triazin-2(1H)-one, 2-Amino-4-hydroxy-1,3,5-triazine
35283847-5G
MOLE35283847-5GEA
0
EUR
MOLE35283847-5G
5-Azacytosin
5-Azacytosin
Formel:
C₃H₄N₄O Molecular Weight: 112,09 g/mol |
MDL:
MFCD00149402 CAS-Nummer: 931-86-2 EINECS: 213-242-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...