Indium(III)trifluormethansulfonat

Lieferant: Molekula

Synonyme: Indium(III) trifluoromethanesulphonate, Indium triflate

35446954-5G
MOLE35446954-5GEA 0 EUR
MOLE35446954-5G
Indium(III)trifluormethansulfonat
Indium(III)trifluormethansulfonat
Formel: C₃F₉InO₉S₃
MDL: MFCD00144478
CAS-Nummer: 128008-30-0
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR