Biphenyl-4-ol
Lieferant: Molekula
Synonyme:
4-Hydroxydiphenyl, 1-Hydroxy-4-phenylbenzol, 4-Hydroxybiphenyl, 4-Phenylphenol
35939801-1KG
MOLE35939801-1KGEA
146
EUR
MOLE35939801-1KG
Biphenyl-4-ol
Biphenyl-4-ol
Formel:
C₁₂H₁₀O Molecular Weight: 170.21 g/mol Siedepunkt: 321 Schmelzpunkt: 162 Dichte: 1.29 g/cm³ (20 °C) |
MDL:
MFCD00002347 CAS-Nummer: 92-69-3 EINECS: 202-179-2 Merck Index: 12,07459 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...