3-Nitro-4-picolin

Lieferant: Molekula

Synonyme: 4-Methyl-3-nitropyridine

36067304-5G
MOLE36067304-5GEA 0 EUR
MOLE36067304-5G
3-Nitro-4-picolin
3-Nitro-4-picolin
Formel: C₆H₆N₂O₂
Molecular Weight: 138,13 g/mol
MDL: MFCD00051829
CAS-Nummer: 5832-44-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR