Methyl-2-brom-5-nitrobenzoat
Lieferant: Molekula
Synonyme:
2-Brom-5-nitrobenzoesäuremethylester
36067944-5G
MOLE36067944-5GEA
0
EUR
MOLE36067944-5G
Methyl-2-brom-5-nitrobenzoat
Methyl-2-brom-5-nitrobenzoat
Formel:
C₈H₆BrNO₄ Molecular Weight: 260,04 g/mol Schmelzpunkt: 82 °C |
MDL:
MFCD00010867 CAS-Nummer: 6942-36-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...