4-Hydroxycumarin
Lieferant: Molekula
Synonyme:
4-Hydroxy-2-benzopyron
36136514-500G
MOLE36136514-500GEA
0
EUR
MOLE36136514-500G
4-Hydroxycumarin
4-Hydroxycumarin
Formel:
C₉H₆O₃ Molecular Weight: 162,14 g/mol Siedepunkt: 349 °C (1013 hPa) Schmelzpunkt: 211…214 °C |
MDL:
MFCD00006856 CAS-Nummer: 1076-38-6 EINECS: 214-060-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...