1,5-Bis(diphenylphosphino)pentan
Lieferant: Molekula
Synonyme:
Pentan-1,3-diylbis(diphenylphosphin), Bis(1,2-diphenylphosphino)pentan, Pentylenbis(diphenylphosphin)
36583793-1G
MOLE36583793-1GEA
0
EUR
MOLE36583793-1G
1,5-Bis(diphenylphosphino)pentan
1,5-Bis(diphenylphosphino)pentan
Formel:
C₂₉H₃₀P₂ Molecular Weight: 440,5 g/mol Siedepunkt: 230 °C (0,1 mmHg) Schmelzpunkt: 43…46 °C |
MDL:
MFCD00003052 CAS-Nummer: 27721-02-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...