Ethyl-2-oxocyclohexancarboxylat
Lieferant: Molekula
Synonyme:
Ethyl 2-oxocyclohexanecarboxylate
36646345-100G
MOLE36646345-100GEA
0
EUR
MOLE36646345-100G
Ethyl-2-oxocyclohexancarboxylat
Ethyl-2-oxocyclohexancarboxylat
Formel:
C₉H₁₄O₃ Molecular Weight: 170,21 g/mol |
MDL:
MFCD00001631 CAS-Nummer: 1655-07-8 EINECS: 216-731-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...