2,5-Dimethylpyrrol

Lieferant: Molekula

Synonyme: 2,5-Dimethyl-1H-pyrrole

36723681-25G
MOLE36723681-25GEA 0 EUR
MOLE36723681-25G
2,5-Dimethylpyrrol
2,5-Dimethylpyrrole
Formel: C₆H₉N
Molecular Weight: 95,1424 g/mol
MDL: MFCD00005223
CAS-Nummer: 625-84-3
EINECS: 210-913-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR