(R)-(-)-1-Phenyl-1,2-ethandiol

Lieferant: Molekula

Synonyme: (R)-(-)-Phenylethan-1,2-diol, (R)-(-)-α,β-Dihydroxyethylbenzol, (R)-(-)-Phenylethylenglycol, (R)-(-)-(1,2-Dihydroxyethyl)benzol

36796198-1G
MOLE36796198-1GEA 0 EUR
MOLE36796198-1G
(R)-(-)-1-Phenyl-1,2-ethandiol
(R)-(-)-1-Phenyl-1,2-ethandiol
Formel: C₈H₁₀O₂
Molecular Weight: 138,17 g/mol
Siedepunkt: 272 °C (1013 hPa)
Schmelzpunkt: 67…69 °C
MDL: MFCD00064262
CAS-Nummer: 16355-00-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR