2,5-Diaminotoluolsulfat

Lieferant: Molekula

37349636-500G
MOLE37349636-500GEA 0 EUR
MOLE37349636-500G
2,5-Diaminotoluolsulfat
2,5-Diaminotoluolsulfat
Formel: C₇H₁₂N₂O₄S
MDL: MFCD00013003
CAS-Nummer: 615-50-9
EINECS: 210-431-8
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR