1,3-Dinitro-4-(trifluormethoxy)benzol

Lieferant: Molekula

Synonyme: 2,4-Dinitro(trifluormethoxy)benzol

37605955-25G
MOLE37605955-25GEA 0 EUR
MOLE37605955-25G
1,3-Dinitro-4-(trifluormethoxy)benzol
1,3-Dinitro-4-(trifluoromethoxy)benzene
Formel: C₇H₃F₃N₂O₅
Molecular Weight: 252,11 g/mol
MDL: MFCD00042268
CAS-Nummer: 655-07-2
EINECS: 470-480-8
UN: 2927
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR