1-(4-Hydroxyphenyl)-5-mercapto-1H-tetrazol

Lieferant: Molekula

Synonyme: 1-(4-Hydroxyphenyl)-5-mercaptotetrazole

37688754-25G
MOLE37688754-25GEA 0 EUR
MOLE37688754-25G
1-(4-Hydroxyphenyl)-5-mercapto-1H-tetrazol
1-(4-Hydroxyphenyl)-5-mercapto-1H-tetrazole
Formel: C₇H₆N₄OS
Molecular Weight: 194,21 g/mol
MDL: MFCD00132898
CAS-Nummer: 52431-78-4
EINECS: 257-909-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR