6-Methoxy-2-naphthol

Lieferant: Molekula

Synonyme: 6-Methoxynaphthalen-2-ol, Naproxen Impurity H

37783588-25G
MOLE37783588-25GEA 785 EUR
MOLE37783588-25G
6-Methoxy-2-naphthol
6-Methoxy-2-naphthol
Formel: C₁₁H₁₀O₂
Molecular Weight: 174.2000 g/mol
MDL: MFCD00088677
CAS-Nummer: 5111-66-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR