Hesperetin
Lieferant: Molekula
Synonyme:
3',5,7-Trihydroxy-4'-methoxyflavanone, 4'-Methoxy-3',5,7-trihydroxyflavanone, (S)-5,7-Dihydroxy-2-(3-hydroxy-4-methoxyphenyl)chroman-4-one, (±)-Hesperetin
37798797-5G
MOLE37798797-5GEA
172
EUR
MOLE37798797-5G
Hesperetin
Hesperetin
Formel:
C₁₆H₁₄O₆ Molecular Weight: 302,30 g/mol |
MDL:
MFCD00075646 CAS-Nummer: 520-33-2 EINECS: 208-290-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...