5-Ethyluracil

Lieferant: Molekula

Synonyme: Ethyluracil, 2,4-Dihydroxy-5-ethylpyrimidin

38127331-1G
MOLE38127331-1GEA 71.6 EUR
MOLE38127331-1G
5-Ethyluracil
5-Ethyluracil
Formel: C₆H₈N₂O₂
Molecular Weight: 140,14 g/mol
MDL: MFCD00079187
CAS-Nummer: 4212-49-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR