5-Methoxy-1-indanon
Lieferant: Molekula
38224728-5G
MOLE38224728-5GEA
134
EUR
MOLE38224728-5G
5-Methoxy-1-indanon
5-Methoxy-1-indanon
Formel:
C₁₀H₁₀O₂ Molecular Weight: 162,19 g/mol Siedepunkt: 145…150 °C (0,2 mmHg) Schmelzpunkt: 107…108 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003789 CAS-Nummer: 5111-70-6 EINECS: 225-838-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...