Triphenylmethan
Lieferant: Molekula
Synonyme:
Tritan
38673033-100G
MOLE38673033-100GEA
0
EUR
MOLE38673033-100G
Triphenylmethan
Triphenylmethan
Formel:
C₁₉H₁₆ Molecular Weight: 244.34 g/mol Siedepunkt: 359 Schmelzpunkt: 90…92 °C Dichte: 1.01 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004763 CAS-Nummer: 519-73-3 EINECS: 208-275-0 Merck Index: 12,09871 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...