(±)-2-Phenoxypropionsäure

Lieferant: Molekula

Synonyme: 2-Phenoxypropanoic acid, 2-Phenoxypropionic acid, alpha-Methylphenoxyacetic acid

38762652-100G
MOLE38762652-100GEA 0 EUR
MOLE38762652-100G
(±)-2-Phenoxypropionsäure
(±)-2-Phenoxypropionsäure
Formel: C₉H₁₀O₃
Molecular Weight: 166,18 g/mol
MDL: MFCD00002643
CAS-Nummer: 940-31-8
EINECS: 213-370-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR