2-Cyanbenzoesäure

Lieferant: Molekula

38822788-1G
MOLE38822788-1GEA 0 EUR
MOLE38822788-1G
2-Cyanbenzoesäure
2-Cyanbenzoesäure
Formel: C₈H₅NO₂
Molecular Weight: 147.13 g/mol
MDL: MFCD00045796
CAS-Nummer: 3839-22-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR